missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Renin Antibody (1), CoraFluor™ 1, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-90510CL1
This item is not returnable.
View return policy
Description
Renin Monoclonal antibody specifically detects Renin in Mouse samples. It is validated for ELISA,Immunohistochemistry (Paraffin)
Specifications
| Renin | |
| Monoclonal | |
| CoraFluor 1 | |
| angiotensin-forming enzyme, Angiotensinogenase, EC 3.4.23, EC 3.4.23.15, FLJ10761, HNFJ2, Prorenin, renin, renin precursor, renal | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Renin (Accession#: NP_112469.1; Met 1-Arg 402). | |
| 0.1 mL | |
| Cardiovascular Biology, metabolism | |
| 5972 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
| ELISA, Immunohistochemistry (Paraffin) | |
| 1 | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction