Learn More
Invitrogen™ Renin 1 (REN1) Mouse ELISA Kit
Sandwich ELISA Kit
Brand: Invitrogen™ EMREN1X5
Additional Details : Weight : 0.33800kg
Description
The Mouse Renin 1 (REN1) ELISA quantitates Ms Renin 1 in mouse serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Ms Renin 1. Principle of the method The Mouse Renin 1 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
Renin is a highly specific endopeptidase, whose only known function is to generate angiotensin I from angiotensinogen in the plasma, initiating a cascade of reactions that produce an elevation of blood pressure and increased sodium retention by the kidney.Specifications
P06281 | |
6 pg/mL | |
Solid-phase sandwich ELISA | |
ELISA Kit | |
Plasma, Serum, Supernatant | |
5 x 96 assays | |
ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis | |
19701 | |
<12% | |
Standard, Adhesive Plate Covers, Wash Buffer, Assay Diluent Concentrate, Chromogen, SAV-HRP, Stop Solution, Biotinylated Detection Antibody, Pre-coated 96 well plates | |
5 x 96 Tests | |
Neurobiology | |
2°C to 8°C | |
Mouse | |
4 hr. 45 min. |
12.21 pg/mL to 50,000 pg/mL | |
6 pg/mL to 50000 pg/mL | |
Biotin | |
Proteases, Peptidases & Inhibitors | |
This ELISA antibody pair detects mouse Renin. Other species not determined yet | |
Mouse | |
Colorimetric Microplate Reader | |
D19352,Ren,Ren-1,Ren-Ac,Ren1d,Rn-1,Rnr,Ren1 | |
<10% | |
HRP | |
RUO | |
Plasma, 50 μ;L; Serum, 50 μ;L; Supernatant, 100 μ;L | |
D19352, Ren, Ren-1, Ren-Ac, Ren1d, Rn-1, Rnr, Ren1 | |
1 hr. 20 min. |