missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ RENBP Recombinant Protein Antigen

Product Code. 18326820 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18326820 0.1mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18326820 Supplier Novus Biologicals™ Supplier No. NBP180852PEP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly purified. Generating reliable and reproducible results. Applications: Antibody Competition

A recombinant protein antigen with a N-terminal His6-ABP tag corresponding to human RENBP. Source: E.coli Amino Acid Sequence: DMEKERETLQAWKERVGQELDRVVAFWMEHSHDQEHGGFFTCLGREGRVYDDLKYVWLQGRQVWMYCRLYRTFERFRHAQLLDAAKAGGEFLLRYARVAPPGKKCAFVLTRDGRPVKVQRTIFSE The RENBP Recombinant Protein Antigen is derived from E. coli. The RENBP Recombinant Protein Antigen has been validated for the following applications: Antibody Competition.

This is a blocking peptide for NBP1-80852. This is a human recombinant protein expressed in E. coli with a N-terminal His-ABP tag and purified with IMAC chromatography.

TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 5973
Species Human
Purification Method Chromatography
Purity >80%
Concentration 0.5mg/mL
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Formulation PBS and 1M Urea, pH 7.4.
For Use With (Application) Blocking/Neutralizing, Control
Gene Symbol RENBP
Label Type Unlabeled
Molecular Weight (g/mol) 32kDa
Product Type RENBP
Quantity 0.1mL
Regulatory Status RUO
Source E.Coli
Specific Reactivity This recombinant antigen is only intended to be used as a blocking agent to confirm antibody specificity with the corresponding antibody, catalog number NBP1-80852. It is purified by IMAC chromatography, and the expected concentration is greater than 0.5 mg/ml
Immunogen DMEKERETLQAWKERVGQELDRVVAFWMEHSHDQEHGGFFTCLGREGRVYDDLKYVWLQGRQVWMYCRLYRTFERFRHAQLLDAAKAGGEFLLRYARVAPPGKKCAFVLTRDGRPVKVQRTIFSE
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.