missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RENBP Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£155.00 - £364.00
Specifications
| Antigen | RENBP |
|---|---|
| Dilution | Western Blot 1:500-1:1000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
RENBP Polyclonal antibody specifically detects RENBP in Human samples. It is validated for Western BlotSpecifications
| RENBP | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cell Biology | |
| PBS (pH 7.3), 50% glycerol | |
| 5973 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:1000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| AGE, EC 5.1.3.8, GlcNAc 2-epimerase, N-acetyl-D-glucosamine 2-epimerase, N-acylglucosamine 2-epimerase, RBP, renin binding protein, Renin-binding protein, RNBP | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 187-390 of human RENBP (NP_002901.2). EPMAVPMMLLNLVEQLGEADEELAGKYAELGDWCARRILQHVQRDGQAVLENVSEGGKELPGCLGRQQNPGHTLEAGWFLLRHCIRKGDPELRAHVIDKFLLLPFHSGWDPDHGGLFYFQDADNFCPTQLEWAMKLWWPHSEAMIAFLMGYSDSGDPVLLRLFYQVAEYTFRQFRDPEYGEWFGYLSREGKVALSIKGGPFKGC | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title