missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Renalase Antibody (RNLS/1940), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08450B
This item is not returnable.
View return policy
Description
Renalase Monoclonal antibody specifically detects Renalase in Human samples. It is validated for ELISA
Specifications
| Renalase | |
| Monoclonal | |
| Biotin | |
| C10orf59chromosome 10 open reading frame 59, EC 1.4, FLJ11218, MAO-C, Monoamine oxidase-C, RENALASE, renalase, FAD-dependent amine oxidase | |
| Recombinant human Renalase protein fragment (around aa 34-235) (exact sequence is proprietary) (Uniprot: Q5VYX0) | |
| 100 μg | |
| Lipid and Metabolism | |
| 55328 | |
| Store at 4°C in the dark. | |
| IgG2b κ |
| ELISA | |
| RNLS/1940 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction