missing translation for 'onlineSavingsMsg'
Learn More

Renalase Antibody (RNLS/1940), Alexa Fluor™ 405, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP3-08450AF405

Product Code. 18574282

  • £471.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Renalase Monoclonal antibody specifically detects Renalase in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Renalase
Monoclonal
Alexa Fluor 405
C10orf59chromosome 10 open reading frame 59, EC 1.4, FLJ11218, MAO-C, Monoamine oxidase-C, RENALASE, renalase, FAD-dependent amine oxidase
Recombinant human Renalase protein fragment (around aa 34-235) (exact sequence is proprietary) (Uniprot: Q5VYX0)
100 μg
Lipid and Metabolism
55328
Store at 4°C in the dark.
IgG2b κ
ELISA
RNLS/1940
50mM Sodium Borate
Mouse
Protein A or G purified
RUO
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.