missing translation for 'onlineSavingsMsg'
Learn More

Renalase Antibody [mFluor Violet 500 SE], Novus Biologicals™

Product Code. 30735951 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30735951 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30735951 Supplier Novus Biologicals Supplier No. AF5350MFV500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody

Renalase Polyclonal antibody specifically detects Renalase in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Renalase
Applications Western Blot
Classification Polyclonal
Conjugate mFluor Violet 500 SE
Formulation 50mM Sodium Borate
Gene Alias C10orf59chromosome 10 open reading frame 59, EC 1.4, FLJ11218, MAO-C, Monoamine oxidase-C, RENALASE, renalase, FAD-dependent amine oxidase
Host Species Goat
Immunogen E. coli-derived recombinant human Renalase, Ala2-Ile342, Accession # NP_001026879
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 55328
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.