missing translation for 'onlineSavingsMsg'
Learn More

Renal Cell Carcinoma Marker (gp200) Ab-1 Mouse Monoclonal Antibody, Epredia™

Provide accurate, reproducible results with the Epredia™ Renal Cell Carcinoma Marker (gp200) Ab-1, Mouse Monoclonal Antibody.

Brand:  Epredia MS-409-RQ

 View more versions of this product

Product Code. 12691367

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

In normal kidney, gp200 is localized along the brush border of the pars convoluta and pars recta segments of the proximal tubule, as well as focally along the luminal surface of Bowman's capsule adjoining the outgoing proximal tubule. Of other normal tissues examined, the gp200 is also localized along the luminal surfaces of breast lobules and ducts, the luminal surface of the epididymal tubular epithelium, within the cytoplasm of parathyroid parenchymal cells, and focally within the colloid of thyroid follicles. Thirty-one other normal tissues do not express similar or cross-reacting antigens. Reportedly, gp200 is expressed by 93% of primary and 84% of metastatic renal cell carcinomas.

Host Species: Mouse

Clone: PN-15

Isotype: IgG1/κ

Species Reactivity: Human and Rat. Others not known.

Epitope: Carbohydrate domain

Immunogen: Microsomal fraction of human renal cortical tissue homogenate

Molecular Weight: ∽200kDa

Positive Control: Normal kidney or renal cell CA

Cellular Localization: Membrane and cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting (Reduced and nonreduced)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Renal Cell Carcinoma Marker (gp200) Ab-1
Monoclonal
Prediluted
Mouse
12 mL
Cancer and Tumor Biology
Ready-to-Use
Immunohistochemistry (Paraffin), Western Blot
PN-15
Unconjugated
Microsomal fraction of human renal cortical tissue homogenate
IVD
Primary
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.