Learn More
Renal Cell Carcinoma Marker (gp200) Ab-1 Mouse Monoclonal Antibody, Epredia™
Provide accurate, reproducible results with the Epredia™ Renal Cell Carcinoma Marker (gp200) Ab-1, Mouse Monoclonal Antibody.
Brand: Epredia MS-409-B0
Description
In normal kidney, gp200 is localized along the brush border of the pars convoluta and pars recta segments of the proximal tubule, as well as focally along the luminal surface of Bowman's capsule adjoining the outgoing proximal tubule. Of other normal tissues examined, the gp200 is also localized along the luminal surfaces of breast lobules and ducts, the luminal surface of the epididymal tubular epithelium, within the cytoplasm of parathyroid parenchymal cells, and focally within the colloid of thyroid follicles. Thirty-one other normal tissues do not express similar or cross-reacting antigens. Reportedly, gp200 is expressed by 93% of primary and 84% of metastatic renal cell carcinomas.Host Species: Mouse
Clone: PN-15
Isotype: IgG1/κ
Species Reactivity: Human and Rat. Others not known.
Epitope: Carbohydrate domain
Immunogen: Microsomal fraction of human renal cortical tissue homogenate
Molecular Weight: ∽200kDa
Positive Control: Normal kidney or renal cell CA
Cellular Localization: Membrane and cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting (Reduced and nonreduced)
Specifications
| Renal Cell Carcinoma Marker (gp200) Ab-1 | |
| Monoclonal | |
| 200μg/mL | |
| Mouse | |
| 100 μL | |
| Cancer and Tumor Biology | |
| Human, Rat | |
| IgG2a κ |
| Immunohistochemistry (Paraffin), Western Blot | |
| PN-15 | |
| Biotin | |
| Microsomal fraction of human renal cortical tissue homogenate | |
| IVD | |
| Primary | |
| Concentrated |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.