Learn More
REN Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 29330-1-AP-150UL
This item is not returnable.
View return policy
Description
The REN antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human REN. This antibody recognizes human, monkey antigen. The REN antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| REN | |
| 500 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Monkey | |
| 150 kDa | |
| IgG |
| Western Blot, ELISA | |
| Angiotensinogenase, FLJ10761, REN, renin | |
| 150 μL | |
| 5972 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |