missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Remoxipride hydrochloride, Tocris Bioscience™
Selective D2-like antagonist
Chemical Identifiers
CAS | 73220-03-8 |
---|---|
Molecular Formula | C16H24BrClN2O3 |
Molecular Weight (g/mol) | 407.733 |
InChI Key | WCPXLMIPGMFZMY-MERQFXBCSA-N |
Synonym | remoxipride hydrochloride, roxiam, remoxipride hcl, a 33547 hydrochloride, unii-qs4s72u30k, s-remoxipride, remoxipride hydrochloride anhydrous, s---2-3-bromo-2,6-dimethoxybenzamido methyl-1-ethylpyrrolidine hydrochloride, s---3-bromo-2,6-dimethoxy-n-1-ethyl-2-pyrrolidinylmethyl benzamide hydrochloride, benzamide, 3-bromo-2,6-dimethoxy-n-1-ethyl-2-pyrrolidinylmethyl-, hydrochloride, s-- |
PubChem CID | 15565709 |
IUPAC Name | 3-bromo-N-[[(2S)-1-ethylpyrrolidin-2-yl]methyl]-2,6-dimethoxybenzamide;hydrochloride |
SMILES | CCN1CCCC1CNC(=O)C2=C(C=CC(=C2OC)Br)OC.Cl |
Description
Dopamine D2 receptor antagonist showing selectivity over D3 and D4 receptors (Ki values are ∽ 300, ∽ 1600, and ∽ 2800 nM for D2, D3 and D4 receptors respectively). Exhibits antipsychotic activity in vivo with no extrapyramidal side effects. 50-fold more potent than sulpiride in antagonising the effects of apomorphine in the rat.Chemical Identifiers
73220-03-8 | |
407.733 | |
remoxipride hydrochloride, roxiam, remoxipride hcl, a 33547 hydrochloride, unii-qs4s72u30k, s-remoxipride, remoxipride hydrochloride anhydrous, s---2-3-bromo-2,6-dimethoxybenzamido methyl-1-ethylpyrrolidine hydrochloride, s---3-bromo-2,6-dimethoxy-n-1-ethyl-2-pyrrolidinylmethyl benzamide hydrochloride, benzamide, 3-bromo-2,6-dimethoxy-n-1-ethyl-2-pyrrolidinylmethyl-, hydrochloride, s-- | |
3-bromo-N-[[(2S)-1-ethylpyrrolidin-2-yl]methyl]-2,6-dimethoxybenzamide;hydrochloride |
C16H24BrClN2O3 | |
WCPXLMIPGMFZMY-MERQFXBCSA-N | |
15565709 | |
CCN1CCCC1CNC(=O)C2=C(C=CC(=C2OC)Br)OC.Cl |
Specifications
73220-03-8 | |
remoxipride hydrochloride, roxiam, remoxipride hcl, a 33547 hydrochloride, unii-qs4s72u30k, s-remoxipride, remoxipride hydrochloride anhydrous, s---2-3-bromo-2,6-dimethoxybenzamido methyl-1-ethylpyrrolidine hydrochloride, s---3-bromo-2,6-dimethoxy-n-1-ethyl-2-pyrrolidinylmethyl benzamide hydrochloride, benzamide, 3-bromo-2,6-dimethoxy-n-1-ethyl-2-pyrrolidinylmethyl-, hydrochloride, s-- | |
CCN1CCCC1CNC(=O)C2=C(C=CC(=C2OC)Br)OC.Cl | |
407.733 | |
407.73 | |
Remoxipride hydrochloride |
C16H24BrClN2O3 | |
WCPXLMIPGMFZMY-MERQFXBCSA-N | |
3-bromo-N-[[(2S)-1-ethylpyrrolidin-2-yl]methyl]-2,6-dimethoxybenzamide;hydrochloride | |
15565709 | |
>98% |
Safety and Handling
Recommended Storage : Store at Room Temperature