Learn More
Thermo Scientific™ Remote Monitoring Dual Point Calibration, ISO 9001
Product Code. 32018033 Shop All Thermo Scientific Products
Change view
Click to view available options
Duration:
1 Day
Unit Size:
Each
Product Code. Duration Unit Size
32018033 1 Day Each
1 options
Product Code. 32018033 Supplier Thermo Scientific™ Supplier No. CALBCRM2
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Remote monitoring dual point calibration. ISO 9001: Certificate with before and after data. Regulatory requirements: Ideal for organization needing to verify equipment performance to manufacturer’s specifications.
Remote monitoring dual point calibration
  • ISO 9001: Certificate with before and after data.
  • Regulatory requirements: Ideal for organization needing to verify equipment performance to manufacturer's specifications. Uncertainties not required.
  • Included in service delivery: Found as left calibration data; detailed calibration label, If a unit is out of tolerance an adjustment is performed
  • Included on accredited certificate: Serial/asset number; Measurement data; Environmental conditions; Conformity assessment; Reference equipment used
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Remote Monitoring Devices
Duration 1 Day
Product Type Calibration