missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRAND™ Remote Dispensing System and Accessories for Dispensette™ Bottletop Dispensers
Shop All Brand ProductsDescription
- 704261- Remote Dispensing System for Dispensette™. Includes wall mount
- 704267 - 10 meters of filling tube
- 704269 - 1 meter filling tube
Specifications
Specifications
| Product Type | FEP Tubing |
| Autoclavable | Not Autoclavable |
| Color | Translucent |
| Material | FEP |
| For Use With (Application) | Remote dispensing |
| For Use With (Equipment) | Dispensette |
| Sterility | Non-sterile |
| Length (Metric) | 1 m |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?