Learn More
Thermo Scientific™ Wellcolex™ E. coli O157
Product Code. 11424109
Change view
Click to view available options
Quantity:
50 Tests/Kit
Unit Size:
50 tests
Product Code. Quantity Unit Size
11424109 50 Tests/Kit 50 tests
1 options
Product Code. 11424109 Supplier Thermo Scientific™ Supplier No. R30959501
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
Rapidly identify Escherichia coli O157 isolates on laboratory media using Wellcolex™ E. coli O157 latex agglutination test.
Rapidly identify Escherichia coli O157 isolates on laboratory media using Thermo Scientific™ Remel™ Wellcolex™ E. coli O157 latex agglutination test. The inability of most E. coli O157:H7 to ferment sorbitol has been used to differentiate them in the laboratory. MacConkey agar containing sorbitol instead of lactose (SMAC) is often used as a primary screening medium1,2. Testing suspect colonies with Wellcolex* E. coli O157 determines whether the isolate belongs to the O157 serogroup.
Verocytotoxin-producing E. coli O157 are an important cause of diarrhoea, haemorrhagic colitis and haemolytic uraemic syndrome3,4. The pathogenicity of this organism is due to the expression of verocytotoxin (VT1 and/or VT2)5-7. Enteritis due to other serotypes is not unusual but in contrast to the majority of isolates, most E. coli O157:H7 do not ferment sorbitol and this has been used as a differential feature for laboratory identification. Testing suspect colonies with Wellcolex* E. coli O157 helps determine whether the isolate belongs to the O157 serogroup.
The Wellcolex* E. coli O157 Rapid Latex Test is categorised as highly complex under the Clinical Laboratory Improvement Act (CLIA88 : Test System Code 40262; Analyte Code 1604).
  • Ready-to-use: Color coded latex and control reagents
  • Rapid: Results within 30 seconds.
  • Easy-to-interpret: Visible agglutination
  • Less sample required
The Wellcolex* E. coli O157 Test Reagent consists of a buffered suspension of red polystyrene latex particles coated with rabbit IgG antibodies specific for E. coli O157 and 0.05% Bronidox® as preservative. When a drop of the reagent is mixed on a card with a suspension of E. coli O157 organisms, rapid agglutination occurs through the interaction of specific IgG and O157 lipopolysaccharide antigen.
Some faecal coliforms can cause non-specific aggregation of latex particles, particularly when grown on sugar containing media such as MacConkey. Therefore a control latex is provided to assist with the identification of non-specific reactions. The control latex consists of a buffered suspension of red polystyrene latex particles coated with non-immune rabbit IgG and 0.05% Bronidox® as preservative.
The kit is also supplied with Positive and Negative controls. The positive control contains a dilute suspension of heat killed E. coli O157 antigens and 0.05% Bronidox® preservative. The negative control contains a dilute suspension of heat killed E. coli O106 antigens and 0.05% Bronidox® preservative.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
TRUSTED_SUSTAINABILITY
Specifications
Description Wellcolex E. coli O157
Detectable Analytes E. coli Serogroup O157
Type E. coli Testing Kit
Quantity 50 Tests/Kit
Nom du produit
Sélectionnez un problème
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.