missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Wellcogen™ Neisseria meningitidis B/E. coli K1 Rapid Latex Agglutination Test

Product Code. 11474089
Change view
Click to view available options
Quantity:
30 Tests/Kit
Unit Size:
30 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11474089 30 Tests/Kit 30 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11474089 Supplier Thermo Scientific™ Supplier No. R30859502
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Rapidly detect antigens to Neisseria meningitidis group B and E. coli K1 present in human body fluids, blood cultures, or on culture media by latex agglutination.

Wellcogen™ N. meningitidis B/E. coli K1 is a rapid latex test for use in the qualitative detection of antigen from Neisseria meningitidis (meningococcus) group B and Escherichia coli K1, present in body fluids as a consequence of infection. The latex can also be used for the detection of specific antigen in blood culture supernatants and for the identification of meningococcus group B or E. coli K1 cultures. Meningococcus group B antigen and E. coli K1 antigen are immunologically indistinguishable from each other. NOTE: Tests performed directly on clinical specimens are intended for screening purposes and should augment, not replace, culture procedures. Results must be used in conjunction with other data; eg symptoms, results of other tests, clinical impressions etc.

Use this rapid latex test in the qualitative detection of antigen from Neisseria meningitidis (meningococcus) group B and Escherichia coli K1 present in body fluids as a consequence of infection.

The latex can also be used for the detection of specific antigen in blood culture supernatants and for the identification of meningococcus group B or E. coli K1 cultures.

The reagent consists of polystyrene latex particles which have been coated with monoclonal antibodies specific to meningococcus group B antigen. These latex particles agglutinate in the presence of sufficient homologous antigen. N.B. The polysaccharide K1 antigen of E. coli shows structural and immunological identity with meningococcus group B antigen, and the reagent does not distinguish between them. Therefore, the reagent may be used to assist in the diagnosis of meningococcal meningitis and neonatal E. coli meningitis.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C
Description Wellcogen Neisseria meningitidis B/E. coli K1 Test Kit
CE Marker Yes
Detectable Analytes N. meningitidis (Group B) Antigen, E. coli K1 Antigen
Type Neisseria Meningitidis Test Kit
Organism Group Group B
Quantity 30 Tests/Kit
Target Organism Class B/E. Coli
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.