Learn More
Thermo Scientific™ Remel™ Voges-Proskauer B Test
Description
Determine the ability of gram-negative bacilli to produce acetylmethylcarbinol (acetoin) from glucose fermentation using Thermo Scientific™ Remel™ 40% Potassium Hydroxide Reagent for Voges-Proskauer B Test. In 1898, Voges and Proskauer reported production of acetylmethylcarbinol from glucose fermentation by certain bacteria when grown in an appropriate medium such as MR-VP Broth1,2. Later Barret made the VP test more sensitive by using 40% Potassium Hydroxide (VP-B) to the medium for the detection of acetylmethylcarbinol production3.
Specifications
Specifications
| Content And Storage | Store at Room Temperature Until Used |
| Description | Voges-Proskauer B |
| Detectable Analytes | Acetylmethylcarbinol |
| Packaging Type | 25 mL/Bottle |
| Quantity | 25 mL |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.