missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Remel™ Tryptic Soy Broth

Product Code. 11973862
Click to view available options
Quantity:
500 g
Unit Size:
500g
This item is not returnable. View return policy

Product Code. 11973862

Brand: Thermo Scientific™ R455052

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Remel Tryptic Soy Broth (TSB) (Soybean Casein Digest) (Dehydrated) is used for the cultivation of a wide variety of nonfastidious microorganisms.

Remel Tryptic Soy Broth (TSB) (Soybean Casein Digest) (Dehydrated) is used for the cultivation of a wide variety of nonfastidious microorganisms.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Industrial Reference: USP
Description Tryptic Soy Broth (TSB) (Soybean Casein Digest)
Form Powder
Product Type Dehydrated Culture Media
Quantity 500 g

For Laboratory Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.