missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Streptex™ Positive Control

Product Code. 13215589
Change view
Click to view available options
Quantity:
2 mL
Unit Size:
2mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
13215589 2 mL 2mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13215589 Supplier Thermo Scientific™ Supplier No. R30164601
  • ONLINE EXCLUSIVE PRICE
    £70.05
    £68.77 / 2mL
    Save £1.28
    1% Off
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Streptex™ Positive Control is a polyvalent positive control that is used with the streptococcal Grouping system Streptex kit.

Validate your test procedures with Thermo Scientific™ Remel™ Streptex™ Positive Control.

Streptex is a rapid latex test system used to detect and identify the Lancefield Groups of streptococci A, B, C, D, F and G and contains all components necessary for the system.

The test utilizes latex particles sensitized with group-specific antibodies which agglutinate in presence of homologous antigens. Streptex Positive Control reagent is polyvalent positive control supplied as an accessory for the Streptex kit.

Streptococci can be generally classified as alpha, beta, or gamma based on their hemolytic reactions indicated by appearance of zones around colonies growing on blood agar. Beta-hemolytic streptococci are further characterized by Lancefield groups.

Most streptococci possess group-specific antigens which are usually carbohydrate structural components of the cell wall. Lancefield showed that these antigens can be extracted in soluble form and identified by precipitation reactions with homologous antisera1. Polystyrene latex particles coated with group-specific antibodies agglutinate strongly in the presence of homologous antigen and remain in smooth suspension in the absence of homologous antigen. The extraction enzyme used is freeze-dried proteolytic fraction obtained from Streptomyces griseus cultures, containing calcium chloride.

  • Ready to use - Latex suspension is supplied as Ready to use reagent

  • Economical solution - Maximize spending efficiency with the option of purchasing separate kit components

  • Color-coded reagent - Red cap

Streptex Polyvalent Positive Control reagent contains a polyvalent extract of antigens from a representative strain of each streptococcal group A, B, C, D, F and G, supplied in single plastic dropper bottles. The solution contains phosphate buffer pH 7.4 and 0.1% sodium azide as preservative.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Control Sets Positive
Description Streptex™ Positive Control
CE Marker Yes
Detectable Analytes Streptococci (Group A, B, C, D, F, G)
Format Vial
Type Streptococcus Test Kit
Material Polyvalent
Organism Group Group A, B, C, D, F and G
Packaging Type 50 Tests/Kit
Quantity 2 mL
Target Organism Class Streptococcus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.