missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Streptex Latex Group A

Product Code. 11444099
Change view
Click to view available options
Quantity:
50 Tests/Kit
Unit Size:
50 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11444099 50 Tests/Kit 50 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11444099 Supplier Thermo Scientific™ Supplier No. R30950601
  • ONLINE EXCLUSIVE PRICE
    £96.35
    £80.98 / 50 tests
    Save £15.37
    15% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Streptex Latex Group A is a supplementary reagent that is used with the streptococcal grouping system Streptex kit.

Supplement the reagents in Thermo Scientific™ Remel™ Streptex™ Latext Agglutination Kit with Thermo Scientific Remel Streptex Latex Group A. Streptex Latex Agglutination Kit is a rapid latex test used to detect and identify the Lancefield Groups of streptococci A, B, C, D, F and G and contains all components necessary for the system. The test utilizes latex particles sensitized with group-specific antibodies which agglutinate in presence of homologous antigens.

Streptococci can be generally classified as alpha, beta, or gamma based on their hemolytic reactions indicated by appearance of zones around colonies growing on blood agar. Beta-hemolytic streptococci are further characterized by Lancefield groups.

Most Streptococci possess group-specific antigens which are usually carbohydrate structural components of the cell wall. Lancefield showed that these antigens can be extracted in soluble form and identified by precipitation reactions with homologous antisera1. Polystyrene latex particles coated with group-specific antibodies agglutinate strongly in the presence of homologous antigen and remain in smooth suspension in the absence of homologous antigen.

  • Ready to use: Latex suspension is supplied as a ready-to-use reagent.

  • Economical solution: Maximize spending efficiency with the option of purchasing separate kit components.

  • Color-coded reagent: Blue cap.

Streptex Latex Group A reagent contains latex specific for group A, sufficient for up to 50 tests, supplied in single plastic dropper bottles. The polystyrene latex particles coated with purified rabbit antibody to the group A antigen are suspended at a concentration of 0.5% in phosphate buffer (pH 7.4) containing 0.1% sodium azide as a preservative.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Description Streptex and Latex Group A
CE Marker Yes
Detectable Analytes Streptococci (Group A)
Type Streptococcus Test Kit
Organism Group Lancefield A
Quantity 50 Tests/Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.