missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Remel™ Spot Indole Reagent

Product Code. 10484203
Click to view available options
Quantity:
25 mL
Unit Size:
25mL
This item is not returnable. View return policy

Product Code. 10484203

Brand: Thermo Scientific™ R21245

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Determine the ability of microorganisms to split indole from tryptophan molecule by spot test method with Spot Indole Reagent.

Determine the ability of an organism to split indole from the tryptophan molecule with Thermo Scientific™ Remel™ Spot Indole Reagent.In 1963, Vracko and Sherris used Spot Indole Reagent for the separation of Proteus species and Escherechia coli1. In 1969, Lowrance, Reich and Traub found ρ-dimethylaminocinnamaldehyde (DMACA) to be the most sensitive indole reagent which could detect 3 mcg of indole per milliliter of medium2.

Spot Indole Reagent is used to determine the ability of microorganisms to split indole from tryptophan molecule by spot test method. Tryptophanases are intracellular enzymes that mediate indole production by hydrolysing the amino acid, tryptophan. This indole combines with DMACA to form a blue-green compound. This is a condensation product formed by an acid splitting of the protein.Ready to use: Liquid reagent.Easy to interpret: Positive results (blue color) are easy to differentiate from negative results (pink color).Spot Indole Reagent contains 1% w/v ρ-dimethylaminocinnamaldehyde (CAS 6203-18-5) in hydrochloric acid (concentrated) (CAS 7647-01-0) and demineralized water (CAS 7732-18-5) supplied in 25 mL bottles.Not all products are available for sale in all territories. Please inquire.Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
TRUSTED_SUSTAINABILITY

Specifications

Description Spot Indole Reagent
Quantity 25 mL
Form Liquid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.