missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Remel™ PYR Disc with Reagent

Product Code. p-4528733
Click to view available options
Quantity:
100 Tests
25 Tests/Kit
50 Test
Unit Size:
100 tests
50 tests
Pack of 25
This item is not returnable. View return policy

Product Code. 12653558

Brand: Thermo Scientific™ R30854401

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Use for presumptive identification of enterococci, Lancefield group A streptococci, and E. coli based on PYRase production.

Thermo Scientific™ Remel PYR Disk w/Reagent is used for presumptive identification of enterococci, Lancefield group A streptococci, and E. coli based on PYRase production.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C
CE Marker Yes
Quantity 100 Tests
compliance-icons
Signal Word
  • Danger
Hazard Category
  • Reproductive toxicity Category 1B
Hazard Statement
  • H360FD-May damage fertility. May damage the unborn child.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.