missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Remel™ PYR Disc with Reagent
Click to view available options
Quantity:
100 Tests
25 Tests/Kit
50 Test
Unit Size:
100 tests
50 tests
Pack of 25
Description
Thermo Scientific™ Remel PYR Disk w/Reagent is used for presumptive identification of enterococci, Lancefield group A streptococci, and E. coli based on PYRase production.
Specifications
Specifications
| Content And Storage | 2°C to 8°C |
| CE Marker | Yes |
| Quantity | 50 Test |
Safety and Handling
Signal Word
- Danger
- Reproductive toxicity Category 1B
- H360FD-May damage fertility. May damage the unborn child.
- P201-Obtain special instructions before use.
- P202-Do not handle until all safety precautions have been read and understood.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction