Learn More
Thermo Scientific™ Positive Control Antisera, Febrile Positive Control
Use as a positive control in rapid screening and tube agglutination tests for the Remel Stained Suspension range.
Brand:  Thermo Scientific™ R30165301
Product Code. 11984092
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
Validate your test procedures with Thermo Scientific™ Remel™ Positive Control Antisera, Febrile Positive Control as a positive control in rapid screening and tube agglutination tests for the Remel Stained Salmonella Suspension range (SS01 to 04, SS09 and SS11 to 13). Remel Stained Salmonella Suspensions are used in the investigation of enteric infections and pyrexia.
Serological tests are based on the fact that antibodies produced in the serum in response to exposure to bacterial antigens will visibly agglutinate with bacteria carrying homologous antigens. Positive Control Antisera, Febrile Positive Control can be used as a positive control in conjunction with the Remel range of Salmonella Stained Suspensions (SS01 to 04, SS09 and SS11 to 13).
  • Ready to use - Prepared reagent
  • Easy to store - Store at 2-8°C until use
  • Easy to interpret -
    • Positive reaction with Salmonella H and O-stained suspensions (SS01 to 04, SS09 and SS11 to 13).
    • Visible agglutination with appropriate bacterial suspensions.
Positive Control Antisera, Febrile Positive Control is produced using rabbit serum and preserved with 0.5% phenol. Each bottle contains 2 mL liquid and is supplied ready to use.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
2°C to 8°C
Yes
2 mL
Febrile Positive Control
2 mL/Vial