missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ PHA Purified

Product Code. 10082333
Change view
Click to view available options
Quantity:
1 Vial
Unit Size:
2mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10082333 1 Vial 2mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10082333 Supplier Thermo Scientific™ Supplier No. R30852801
  • ONLINE EXCLUSIVE PRICE
    £709.13
    £321.92 / 2mg
    Save £387.21
    54% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Phytohaemagglutinin (PHA) is a mitogenic agent which is used in studies involving cell permeability.

Carry out studies involving cell permeability with Thermo Scientific™ Remel™ Phytohaemagglutinin (PHA). PHA is a mitogenic agent derived from extracts of Phaseolus vulgaris seeds and has twin properties of causing erythroagglutination and of stimulating progressive lymphocyte mitosis in cell culture. PHA has proved to be beneficial in the study of the immune response, lymphocyte kinetics, and bone marrow dynamics, and for these purposes it is desirable to use a substance which has more closely reproducible qualities and known potency.

Purified phytohaemagglutinin is a freeze-dried, highly refined protein fraction of selected Phaseolus spp. seed extract in which the specific mitogenic activity and the mitogenic/ haemagglutinating activity ratio have been increased by a factor of about 100/1 during purification. Both agglutinating and mitogenic activities of PHA are found to be associated with the protein fractions of crude extracts, which are sufficiently alike in their physico-chemical properties, thus it is difficult to separate them. Relatively crude extracts selected for their ability to yield good mitotic pictures have proved to be useful in routine lymphocyte culture for chromosome studies.

  • Ready to use - Prepared reagent
  • Easy to store - Store at 2° to 8°C until use
  • Can be used in routine lymphocyte culture for chromosome studies

Purified phytohaemagglutinin contains a freeze-dried, highly refined protein fraction of selected Phaseolus spp. seed extract. Each bottle contains 2mg lyophilised phytohaemagglutinin (purified), dried from a small volume of buffered saline without preservative. The reagent may be reconstituted to any desired volume in sterile saline (if buffered, preferably to a pH between 6.5 and 8) or distilled water, using a sterile disposable hypodermic syringe.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C
Description PHA Purified
Format Vial
Quantity 1 Vial
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.