missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Remel™ PathoDxtra™ Strep Grouping Kit

Product Code. 12896333
Click to view available options
Quantity:
1 Set
2.8 mL
50 Cards
60 Tests
60 Tests/Kit
Description:
Disposable Reaction Cards
Polyvalent Positive Control
Strep Group A Latex
Strep Group B Latex
Strep Group C Latex
Strep Group D Latex
Strep Group F Latex
Strep Group G Latex
Strep Grouping Kit
Strep Grouping Reagent Set
Unit Size:
3mL
60 tests
Pack of 50
This item is not returnable. View return policy

Product Code. 12896333

Brand: Thermo Scientific™ DR0707G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Eliminates need for incubation and provides a faster strep grouping solution than more traditional enzyme extraction.

Eliminate the need for incubation and provide a faster strep grouping solution than more traditional enzyme extraction with the Thermo Scientific™ PathoDxtra™ Strep Grouping Kit.

  • Improves workflow efficiencies — Instant acid extraction without the need for incubation or reconstitution
  • Saves time — Simple, rapid procedure produces easy-to-read results
  • Reduces costs — Identifies all clinically important Strep groups, including Group D; no additional reagents required
  • Provides flexibility — Pack variants available to suit testing needs

Includes:

PathoDxtra Strep Grouping Kit (Mfr. No. DR0700M): 3mL each of A, B, C, D, F and G grouping latex; Polyvalent Positive Control, 2.8mL; Extraction Reagent 1: 4mL; Extraction Reagent 2: 4mL; Extraction Reagent 3: 2 x 10mL; Mixing Sticks; Disposable Reaction Cards

PathoDxtra Extraction Reagent Set (Mfr. No. DR0709M): Extraction Reagent 1: 4mL; Extraction Reagent 2: 4mL; Extraction Reagent 3: 2 x 10mL; For use with the PathoDxtra Strep Grouping Kit

PathoDxtra Strep Grouping Reagent Set (Mfr. No. DR0710M): Extraction Reagent 1 - 4mL; Extraction Reagent 2 - 4mL; Extraction Reagent 3 - 2 x 10mL; Mixing Sticks, Disposable Reaction Cards; For use with selected, separately purchased PathoDxtra Strep Grouping latex reagents

TRUSTED_SUSTAINABILITY

Specifications

Control Sets Positive
Description Polyvalent Positive Control
Detectable Analytes beta-haemolytic streptococci
Type Positive Control
For Use With (Application) Enzyme Immunoassay Detection
Material Polyvalent
Organism Group Group A, B, C, D, F and G
Quantity 2.8 mL

Rapid Streptococcal grouping tests correlate best with reference methods when only Streptococci that are Β-hemolytic on sheep blood agar are tested. 1, 2, 3, 4

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.