missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Remel™ PathoDxtra™ Strep Grouping Kit

Product Code. 12886333
Change view
Click to view available options
Quantity:
1 Set
2.8 mL
50 Cards
60 Tests
60 Tests/Kit
Description:
Disposable Reaction Cards
Extraction Reagents
Polyvalent Positive Control
Strep Group A Latex
Strep Group B Latex
Strep Group C Latex
Strep Group D Latex
Strep Group F Latex
Strep Group G Latex
Strep Grouping Kit
Strep Grouping Reagent Set
Unit Size:
3mL
60 tests
Pack of 50
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Description Unit Size
12886333 60 Tests Strep Group G Latex 60 tests
12806333 60 Tests Strep Group D Latex 60 tests
12846333 60 Tests Strep Group F Latex 60 tests
12846323 60 Tests Strep Group A Latex 60 tests
12836353 1 Set Strep Grouping Reagent Set 60 tests
12896333 2.8 mL Polyvalent Positive Control 3mL
12876323 60 Tests Strep Group B Latex 60 tests
12866343 1 Set Extraction Reagents 60 tests
13206059 60 Tests/Kit Strep Grouping Kit 60 tests
12806373 50 Cards Disposable Reaction Cards Pack of 50
12886323 60 Tests Strep Group C Latex 60 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
This item is not returnable. View return policy
Product Code. 12886333 Supplier Thermo Scientific™ Supplier No. DR0706G
  • ONLINE EXCLUSIVE PRICE
    £56.40
    £51.31 / 60 tests
    Save £5.09
    9% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Eliminates need for incubation and provides a faster strep grouping solution than more traditional enzyme extraction.

Eliminate the need for incubation and provide a faster strep grouping solution than more traditional enzyme extraction with the Thermo Scientific™ PathoDxtra™ Strep Grouping Kit.

  • Improves workflow efficiencies — Instant acid extraction without the need for incubation or reconstitution
  • Saves time — Simple, rapid procedure produces easy-to-read results
  • Reduces costs — Identifies all clinically important Strep groups, including Group D; no additional reagents required
  • Provides flexibility — Pack variants available to suit testing needs

Includes:

PathoDxtra Strep Grouping Kit (Mfr. No. DR0700M): 3mL each of A, B, C, D, F and G grouping latex; Polyvalent Positive Control, 2.8mL; Extraction Reagent 1: 4mL; Extraction Reagent 2: 4mL; Extraction Reagent 3: 2 x 10mL; Mixing Sticks; Disposable Reaction Cards

PathoDxtra Extraction Reagent Set (Mfr. No. DR0709M): Extraction Reagent 1: 4mL; Extraction Reagent 2: 4mL; Extraction Reagent 3: 2 x 10mL; For use with the PathoDxtra Strep Grouping Kit

PathoDxtra Strep Grouping Reagent Set (Mfr. No. DR0710M): Extraction Reagent 1 - 4mL; Extraction Reagent 2 - 4mL; Extraction Reagent 3 - 2 x 10mL; Mixing Sticks, Disposable Reaction Cards; For use with selected, separately purchased PathoDxtra Strep Grouping latex reagents

TRUSTED_SUSTAINABILITY

Specifications

Control Sets Positive
Description Strep Group G Latex
Detectable Analytes beta-haemolytic Streptococci
Type Streptococcus Test Kit
For Use With (Application) Enzyme Immunoassay Detection
Material Polyvalent
Organism Group Group A, B, C, D, F and G
Quantity 60 Tests

Rapid Streptococcal grouping tests correlate best with reference methods when only Streptococci that are Β-hemolytic on sheep blood agar are tested. 1, 2, 3, 4

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.