missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Oxichrome Reagent

Product Code. 12958264
Click to view available options
Quantity:
15 mL/Vial
Unit Size:
15mL
This item is not returnable. View return policy

Product Code. 12958264

Brand: Thermo Scientific™ R8309009

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Rapidly detect the enzyme cytochrome oxidase in microorganisms using Oxichrome Reagent.

Rapidly detect the enzyme cytochrome oxidase in microorganisms using Thermo Scientific™ Remel™ Oxichrome Reagent. The oxidase test was originally developed to identify Neisseria spp., but later was used to separate pseudomonads from oxidase-negative enteric Gram-negative bacilli (Enterobacteriaceae)1,2. Most Gram-positive bacteria are oxidase-negative, pseudomonads are oxidase-positive, and other non-enteric, Gram-negative bacilli, are variable

Pseudomonads, bacteria of the family Pseudomonadaceae are a large and varied group comprising four major genera and several hundred species. Most species of the family are found in soil or water; some cause diseases in plants, and a few cause serious diseases in humans and other mammals. Pseudomonas aeruginosa, very common and widespread, is an opportunistic pathogen for man that causes antibiotic-resistive infections in persons of weakened resistance.

The oxidase reaction is attributed to the presence of a cytochrome oxidase system in the respiratory chain of certain bacteria, which utilizes molecular oxygen as a terminal electron acceptor3,4. Oxidase-positive bacteria produce oxidase enzymes which, in the presence of atmospheric oxygen, cytochrome c, and an oxidase reagent (Oxichrome Reagent), oxidize the reagent to form a blue-purple colored compound, Wurster’s blue.

  • Ready-to-use – Liquid reagent
  • Easy to interpret – Blue to purple color indicates a positive test
  • Easy-to-store – Storage at room temperature
  • Rapid results – Can be interpreted within 30 seconds

Oxichrome reagent contains N,N,N,N-tetramethyl-ρ-phenylenediamine supplied in 15 mL bottles.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at Room Temperature Until Used
Description Oxichrome Reagent
Format Vial
Packaging Type 15 mL/Vial
Quantity 15 mL/Vial
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.