Learn More
Thermo Scientific™ Remel™ OADC Enrichment
Description
Remel OADC Enrichment is used in Middlebrook Media.
Specifications
Specifications
| Description | OADC Enrichment |
| Form | Liquid |
| Product Type | OADC Enrichment |
| Quantity | 500 mL |