missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nitrocefin Disk

Product Code. 13265649
Change view
Click to view available options
Quantity:
25 Disks/Vial
Unit Size:
Pack of 25
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
13265649 25 Disks/Vial Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13265649 Supplier Thermo Scientific™ Supplier No. R211667

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Nitrocefin-impregnated disk recommended for use in qualitative procedures for rapid detection of β-lactamase production by certain bacteria.

Detect β-lactamase production by Neisseria gonorrhoeae, Haemophilus influenzae, Moraxella catarrhalis, Staphylococcus spp., Enteroccoccus spp., and anaerobic bacteria with Thermo Scientific™ Remel™ Nitrocefin Disk. The β-lactamase enzyme is produced by various organisms and is responsible for their resistance to penicillins and cephalosporins1,2. Since then, similar enzymes with somewhat different substrate specificities have been identified in various bacterial species. The test methods used to detect β-lactamase include iodometric, acidimetric, and chromogenic procedures3,4.

Since Abraham and Chain confirmed β-lactamase production by microorganisms to be responsible for antibiotic resistance, similar enzymes have been identified in various species. Some enzymes selectively hydrolyze penicillin class antibiotics (i.e., penicillinases), other enzymes hydrolyze all β-lactamase antibiotics except carbapenems (i.e. cephalosporinases) and some enzymes hydrolyze both5.

  • Ready to use - Nitrocefin impregnated disks.
  • Rapid results
  • Conforms to CLSI - Performance Standards for Antimicrobial Disk Susceptibility Tests – (Approved Standard M2-A8 and M11-A5)
  • Easy to interpret - With color change

The cephalosporin nitrocefin is the substrate used in this test. β-lactamase hydrolyzes the β-lactam ring of nitrocefin, producing cephalosporanic acid6. This results in the pale yellow nitrocefin changing to a pink product upon hydrolysis7. Aerobic and anaerobic β-lactamase-producing bacteria affect this color; organisms that fail to produce β-lactamase do not alter the pale-yellow color of nitrocefin within the time limits of test.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C
Description Nitrocefin Discs
CE Marker Yes
Detectable Analytes beta-lactamase (Haemophilus species, N. gonorrhoeae,Staphylococci)
Format Vial
Quantity 25 Disks/Vial
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.