missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Remel™ Ninhydrin Reagent

Product Code. 10586383 Shop All Thermo Scientific Products
Click to view available options
Quantity:
25 mL
Unit Size:
25mL
This item is not returnable. View return policy

Product Code. 10586383

Brand: Thermo Scientific™ R21238

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Detect sodium hippurate hydrolysis by beta-hemolytic streptococci, G. vaginalis, and C. jejuni with with Ninhydrin Reagent.

Detect hydrolysis of sodium hippurate by beta-hemolytic streptococci, Gardnerella vaginalis, and Campylobacter jejuni with Thermo Scientific™ Remel™ Ninhydrin Reagent.The hippurate hydrolysis test determines the ability of group B streptococci as well as other bacteria to enzymatically hydrolyze sodium hippurate1-3. Hwang and Ederer employed the Ninhydrin Reagent in a rapid method to detect hippurate hydrolysis by the end product glycine4.

Ninhydrin Reagent is used to detect sodium hippurate hydrolysis by beta-hemolytic streptococci, G. vaginalis, and C. jejuni.Hippuric acid is hydrolyzed by the enzyme hippuricase to glycine and benzoic acid. Ninhydrin evokes a five-step reaction to form hydrindantin (a reduced form of ninhydrin), carbon dioxide, and ammonia. A condensation reaction happens with hydrindantin, ammonia and residual ninhydrin to produce a final purple colored complex3.Ready for use: no further preparation is required.Easy to interpret:Positive test: Blue to purple color development within 30 minutes.Negative test: Light grey color development or no color change within 30 minutes.Ninhydrin Reagent contains ninhydrin (CAS 485-47-2), acetone (CAS 67-64-1), butanol (CAS 71-36-3) supplied in 25 mL bottles. This product is for in vitro diagnostic use and should be used by properly trained individuals. This reagent have been tested using quality control procedure and found to be acceptable.For use with Hippurate Disk, REF R2108Not all products are available for sale in all territories. Please inquire.Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C
Description Ninhydrin Reagent
CE Marker Yes
Detectable Analytes Glycine, Benzoic Acid
Format Bottle
Quantity 25 mL
compliance-icons compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.