missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Middlebrook 7H9 Broth (Dehydrated)

Product Code. p-6092543
Click to view available options
Quantity:
500 g
Unit Size:
500g
This item is not returnable. View return policy

Product Code. 15228977

Brand: Thermo Scientific™ R454012

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Isolate and cultivate Mycobacterium spp. using Middlebrook 7H9 Broth.

Isolate and cultivate Mycobacterium spp. using Thermo Scientific™ Remel™ Middlebrook 7H9 Broth (Dehydrated). In 1958, Middlebrook and Cohn were the first to formulate this medium for the cultivation of mycobacteria1. This medium is found useful in the transfer of cultures to be tested with biochemical media2.

Isolate and cultivate Mycobacterium spp. using Middlebrook 7H9 Broth.

  • Ease of Handling: Ease of transfer of cultures to be tested with biochemical media.
  • Allows luxuriant growth of Mycobacterium

This medium is used for qualitative procedures. This medium contains inorganic salts which are essential for the growth of mycobacteria. Ammonium sulphate acts as a source of nitrogen. Biotin plays an important role in carboxylation and decarboxylation reactions, while pyridoxin is a precursor of enzymatic activity. The addition of ADC Enrichment provides a carbon source of energy, and catalase minimizes toxic effects.

 

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Description Middlebrook 7H9 Broth
Form Powder
Product Type Dehydrated Culture Media
Quantity 500 g

For Laboratory Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.