missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Microdase Disks

Product Code. 10250483
Change view
Click to view available options
Quantity:
25 Disks/Vial
Unit Size:
Pack of 25
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10250483 25 Disks/Vial Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10250483 Supplier Thermo Scientific™ Supplier No. R21132
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Rapidly differentiate Staphylococcus from Micrococcus species by detecting oxidase enzyme within 2 minutes with this reagent-impregnated disk.

Differentiate Staphylococcus from Micrococcus by detecting the oxidase enzyme with Thermo Scientific™ Remel™ Microdase Disk.

In 1956, Kovacs1 described the oxidase method that can be used to differentiate Gram-negative bacilli. Faller and Schleifer2 modified Kovacs' oxidase reagent by using a tetramethyl-p-phenylenediamine (TMPD) in dimethyl sulfoxide (DMSO). Tarrand and Groschel3 further tested modified Kovacs and confirmed it to be far superior to Kovacs' oxidase reagent.

All micrococci have cytochrome c whereas most staphylococci (with a few exceptions) lack this type of cytochrome. The oxidase reagent confirms the presence of cytochrome c4 by exhibiting color change to blue or purple.

  • Ready to use: Reagent-impregnated disk. No further preparation required.
  • Easy to read: Visible color change that confirms positive test
    Positive test: Blue or purple color development within 2 minutes
    Negative test: No color change (white to grey color remains) within 2 minutes.

Each Microdase Disk is impregnated with tetramethyl-p-phenylenediamine (TMPD) in dimethyl sulfoxide (DMSO). The DMSO provides solubility and stability against auto-oxidation and enhances the permeability of TMPD into the bacterial cell. In presence of atmospheric oxygen, the oxidase enzyme reacts with TMPD and cytochrome c in the organism to form the colored compound indophenol.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C
Description Microdase Discs to differentiate staphylococci from micrococci
CE Marker Yes
Detectable Analytes Oxidase Enzyme
Format Vial
Quantity 25 Disks/Vial
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.