Learn More
Thermo Scientific™ Remel™ McFarland Equivalence Turbidity Standards
Thermo Scientific™ Remel McFarland Equivalence Turbidity Standard is packaged in plastic case with a visual comparison card.
Brand:  Thermo Scientific™ R20415
Code : NEW
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 11454049
  • £81.42 / Each
In Stock
to see stock.
Add to basket
Description
Description
Thermo Scientific™ Remel McFarland Equivalence Turbidity Standard is packaged in plastic case with a visual comparison card.
  • Tube size is 15 x 103mm and fits in most spectrophotometers
  • Polystyrene technology allows longer shelf life
Specifications
Specifications
Turbidity Standard 5.0
15 x 103 mm