Learn More
Thermo Scientific™ Remel™ McFarland Equivalence Turbidity Standards
Thermo Scientific™ Remel McFarland Equivalence Turbidity Standard is packaged in plastic case with a visual comparison card.
Brand:  Thermo Scientific™ R20414
Code : 50
Additional Details : Weight : 0.50000kg
 View more versions of this product
Product Code. 10108582
  • £140.00 / Each
In Stock
to see stock.
Add to basket
Description
Description
Thermo Scientific™ Remel McFarland Equivalence Turbidity Standard is packaged in plastic case with a visual comparison card.
  • Tube size is 15 x 103mm and fits in most spectrophotometers
  • Polystyrene technology allows longer shelf life
Specifications
Specifications
Turbidity Standard 4.0
15 x 103 mm