missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ M Broth (Dehydrated)

Product Code. 11922912
Click to view available options
Quantity:
500 g
Unit Size:
500g
This item is not returnable. View return policy

Product Code. 11922912

Brand: Thermo Scientific™ R453782

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Detect and cultivate of Salmonella in dried foods and feeds using M Broth.

Detect and cultivate Salmonella in dried foods and feeds using Thermo Scientific™ Remel™ M Broth (Dehydrated). This enrichment medium was first formulated in 1969 by Sperber and Deibel using an accelerated serology technique1. This media increases rapid growth of Salmonella, improve flagella development and prevents nonspecific agglutination of bacteria cells used in serology techniques. It is currently recommended by the AOAC International Official Methods of Analysis, and Microbiology Laboratory Guidebook for the detection of Salmonella in foods2-4.

Accelerated detection and cultivation of Salmonella in dried foods and feeds using M Broth.

  • Accelerated Serology Technique: For detection of Salmonella in dried foods and feeds which prevents delayed transportation of bulk feeds to the market place unlike previous methods.
  • Recommended by the AOAC International Official Methods of Analysis, and Microbiology Laboratory Guidebook for the detection of Salmonella in foods2-4.

This medium is used for qualitative procedures. This medium contains casein peptone that provides amino acids and nitrogenous compounds necessary for the bacterial growth and yeast extract that provides B-complex vitamins. Mannose is the carbon source and serves to prevent fimbrial agglutination in serological techniques. Polysorbate 80 acts as a surfactant and dispersing agent.

 

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Description M Broth
Form Powder
Product Type Dehydrated Culture Media
Quantity 500 g

For Laboratory Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.