missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Catarrhalis Test Disk

Product Code. 10516963
Change view
Click to view available options
Quantity:
25 Disks/Vial
Unit Size:
Pack of 25
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10516963 25 Disks/Vial Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10516963 Supplier Thermo Scientific™ Supplier No. R21121
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

Rapidly detect butyrate esterase to presumptively identify Moraxella catarrhalis isolated from clinical specimens with this ready-to-use, reagent-impregnated disk.

Detect butyrate esterase to rapidly identify Moraxella catarrhalis isolated from clinical specimens with Thermo Scientific™ Remel™ Catarrhalis Test Disk.

The rapid identification of Moraxella catarrhalis is important, since most strains produce beta-lactamase and are resistant to penicillin and ampicillin. Conventional methods used to identify M. catarrhalis may take up to 48 hours to perform3,4. In 1962, Berger suggested that detection of butyrate esterase can be used as a rapid method to identify M. catarrhalis.1

Detect butyrate esterase to presumptively identify Moraxella catarrhalis using the reagent-impregnated Catarrhalis Test Disk. Moraxella catarrhalis is an important pathogen. Yeun et al. have reported that the isolation of M. catarrhalis from routine sputum cultures is associated with clinical infection.2 Rapid identification of this organism is important, since most strains produce beta-lactamase and are resistant to penicillin and ampicillin.

  • Ready to use: Reagent-impregnated disk. No further preparation required.
  • Easy to detect: Visual detection of color change.
  • Easy to interpret:
    • Positive test- blue-green color development within 2 minutes
    • Negetive test- no color development within 2 minutes.

Catarrhalis Test Disk is impregnated with indoxyl butyrate as the reactive ingredient. The enzyme butyrate esterase releases indoxyl from indoxyl butyrate and spontaneously forms indigo in the presence of oxygen5. This color change allows rapid detection of M. catarrhalis.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C
Description Detection of butyrate esterase for presumptive identification of Moraxella catarrhalis
CE Marker Yes
Detectable Analytes Butyrate esterase
Format Vial
Quantity 25 Disks/Vial
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.