missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Remel™ BHI Broth

Product Code. 11982902
Click to view available options
Quantity:
500 g
Unit Size:
500g
This item is not returnable. View return policy

Product Code. 11982902

Brand: Thermo Scientific™ R452472

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Cultivate a wide variety of fastidious and non-fastidious microorganisms using BHI Broth (Dehydrated).

Cultivate a wide variety of fastidious and non-fastidious microorganisms using Thermo Scientific™ Remel™ BHI Broth (Dehydrated). Rosenow developed a medium for culturing Streptococci by adding pieces of brain tissue to dextrose broth1. BHI Broth is used to cultivate a wide variety of microorganisms including bacteria, yeasts, and molds. It is also used for the preparation of inocula for use in antimicrobial susceptibility tests and broth microdilution for minimum inhibitory concentration (MIC) testing. This medium has been recommended by the Food and Drug Administration2.

This medium contains peptones and infusions of brain and heart tissue which supply protein and other nutrients necessary for the growth of bacteria. Sodium chloride provides essential electrolytes and maintains osmotic equilibrium. Disodium phosphate acts as a buffer and dextrose is a source of energy.

TRUSTED_SUSTAINABILITY

Specifications

Description BH1 Broth
Form Powder
Product Type Dehydrated Culture Media
Quantity 500 g

For Laboratory Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.