missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Remel™ BactiDrop™ Potassium Hydroxide

Product Code. 13265819
Click to view available options
Quantity:
50 x 0.75 mL
Unit Size:
Pack of 50
This item is not returnable. View return policy

Product Code. 13265819

Brand: Thermo Scientific™ R21524

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Prepare clinical specimens for microscopic examination of fungal elements with BactiDrop™ Potassium Hydroxide.

Prepare clinical specimens for microscopic examination of fungal elements with Thermo Scientific™ Remel™ BactiDrop™ Potassium Hydroxide.The Laboratory Handbook of Medical Mycology recommends the use of 10% KOH for rapid detection of fungal elements1. McGowan reported the use of 10% KOH but warned that a lack of contrast between fungal elements and clinical specimens may occur2. Hence, she recommended phase-contrast microscopy for improved visibility. The formulation of 10% KOH is recommended by the Mayo Clinic Mycology Laboratory3.

Potassium Hydroxide (KOH) is used to prepare clinical specimens for microscopic examination of fungal elements. KOH preparations may be made for all clinical specimens except spinal fluid and urine4. KOH disrupts cellular sheets or clumps of proteinaceous debris that may be present and clears the specimen which makes it easy to detect fungus5. Glycerin in the KOH solution reduces the amount of precipitation in the reagent container and prevents KOH preparations from drying for at least 18-24 hours.Potassium Hydroxide contains potassium hydroxide (CAS 1310-58-3), glycerine (CAS 56-81-5) and demineralized water (CAS 7732-18-5). This product is for in vitro diagnostic use and should be used by properly trained individuals. This reagent has been tested using quality control procedure and found to be acceptable. The reagent is supplied as 0.75 mL ampules.Not all products are available for sale in all territories. Please inquire.Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 20°C to 25°C
Description BactiDrop Potassium Hydroxide
CE Marker Yes
Detectable Analytes Fungus
Quantity 50 x 0.75 mL
compliance-icons
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.