missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Remel™ BactiCard™ Strep

Product Code. 13265659
Change view
Click to view available options
Quantity:
25 Tests/Kit
Unit Size:
25 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
13265659 25 Tests/Kit 25 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13265659 Supplier Thermo Scientific™ Supplier No. R21112
  • ONLINE EXCLUSIVE PRICE
    £295.00
    £268.17 / 25 tests
    Save £26.83
    9% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Thermo Scientific™ Remel BactiCard Strep is used for presumptive identification of streptococci and related genera using enzyme technology.

Thermo Scientific™ Remel BactiCard Strep is used for presumptive identification of streptococci and related genera using enzyme technology.

TRUSTED_SUSTAINABILITY

Specifications

Control Sets Positive, Negative
Description BactiCard™ Strep
CE Marker Yes
Detectable Analytes Clumping Factor and Protein A
Type Bacticard for Streptococcus Testing
For Use With (Application) Enzyme Immunoassay Detection
Packaging Type 1 Kit
Quantity 25 Tests/Kit
Target Organism Class Staphylococcus aureus
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.