missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Agglutinating Sera, Salmonella O

Product Code. 11964092
Change view
Click to view available options
Description:
Factor 10 (group E1)
Factor 11 (group F)
Factor 13, 22 (group G)
Factor 14 (group H)
Factor 15 (group E2)
Factor 16 (group I)
Factor 17 (group J)
Factor 19 (group E4)
Factor 2 (group A)
Factor 27 (group B)
Factor 39 (group Q)
Factor 4 (group B )
Factor 5 (group B)
Factor 6, 7 (group C1)
Factor 8 (group C2)
Factor 9 (group D)
Factors 3, 10, 15, 19 (group E)
Unit Size:
2mL
Each
17 product options available for selection
Product selection table with 17 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Description Unit Size
11964092 Factor 4 (group B ) 2mL
11933332 Factors 3, 10, 15, 19 (group E) 2mL
12653638 Factor 16 (group I) Each
12697559 Factor 11 (group F) Each
11963332 Factor 2 (group A) 2mL
11913332 Factor 6, 7 (group C1) 2mL
12663648 Factor 39 (group Q) Each
11954092 Factor 8 (group C2) 2mL
12697569 Factor 27 (group B) Each
12687559 Factor 10 (group E1) Each
12623638 Factor 13, 22 (group G) Each
12643638 Factor 15 (group E2) Each
12633638 Factor 14 (group H) Each
11934092 Factor 5 (group B) 2mL
15294927 Factor 17 (group J) -
12683638 Factor 19 (group E4) Each
11944092 Factor 9 (group D) 2mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 17 options available.
17 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 11964092 missing translation for 'mfr' Thermo Scientific™ missing translation for 'supplierNo' R30956901
  • ONLINE EXCLUSIVE PRICE
    £139.00
    £126.62 / 2mL
    Save £12.38
    8% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Serologically identify somatic O antigens of Salmonella strains by slide agglutination test using Agglutinating Sera, Salmonella O.

Serologically identify somatic O antigens of Salmonella strains by slide agglutination test for epidemiological and diagnostic purposes using Thermo Scientific™ Remel™ Agglutinating Sera, Salmonella O. The sera are absorbed to remove agglutinins for other Salmonella antigens and the paracolon α agglutinin. Salmonella Somatic Agglutinating Sera (ZC series) are intended for the identification of O antigens.

  • Select sera also for use as positive controls for stained Salmonella suspensions
TRUSTED_SUSTAINABILITY

Specifications

Control Sets Positive
Description Factor 4 (group B )
Detectable Analytes Salmonella O Antigen
Type Agglutinating Sera
Organism Group Group B
Packaging Type 2 mL/Vial
Quantity 2 mL
Target Organism Class Salmonella O
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.