missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Agglutinating Sera, Salmonella O

Product Code. 11934092
Click to view available options
Description:
Factor 10 (group E1)
Factor 11 (group F)
Factor 13, 22 (group G)
Factor 14 (group H)
Factor 15 (group E2)
Factor 16 (group I)
Factor 17 (group J)
Factor 19 (group E4)
Factor 2 (group A)
Factor 27 (group B)
Factor 39 (group Q)
Factor 4 (group B )
Factor 5 (group B)
Factor 6, 7 (group C1)
Factor 8 (group C2)
Factor 9 (group D)
Factors 3, 10, 15, 19 (group E)
Unit Size:
2mL
Each
This item is not returnable. View return policy

Product Code. 11934092

Brand: Thermo Scientific™ R30957001

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Serologically identify somatic O antigens of Salmonella strains by slide agglutination test using Agglutinating Sera, Salmonella O.

Serologically identify somatic O antigens of Salmonella strains by slide agglutination test for epidemiological and diagnostic purposes using Thermo Scientific™ Remel™ Agglutinating Sera, Salmonella O. The sera are absorbed to remove agglutinins for other Salmonella antigens and the paracolon α agglutinin. Salmonella Somatic Agglutinating Sera (ZC series) are intended for the identification of O antigens.

  • Select sera also for use as positive controls for stained Salmonella suspensions
TRUSTED_SUSTAINABILITY

Specifications

Control Sets Positive
Description Factor 5 (group B)
Detectable Analytes Salmonella O Antigen
Type Agglutinating Sera
Organism Group Group B
Packaging Type 2 mL/Vial
Quantity 2 mL
Target Organism Class Salmonella O
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.