Learn More
Thermo Scientific™ Remel™ Agglutinating Sera, Salmonella H
Used for identification of flagellar H antigens of Salmonella strains by slide and/or tube agglutination test.
Brand: Thermo Scientific™ R30162801
Code : 50
Additional Details : Weight : 0.01000kg
Description
Remel Agglutinating Sera, Salmonella H is used for identification of flagellar H antigens of Salmonella strains by slide and/or tube agglutination test.
- Select sera also for use as positive controls for stained Salmonella suspensions
Specifications
Positive | |
Salmonella H Antigen | |
2 mL/Vial | |
Salmonella H |
Salmonella H (z27) | |
Agglutinating Sera | |
2 mL |