missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Remel™ Agglutinating Sera, Neisseria meningitidis

Product Code. 13295589
Change view
Click to view available options
Description:
Group A
Group B
Group C
Group D
Group W135
Unit Size:
2mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Description Unit Size
13295589 Group W135 2mL
13255589 Group C 2mL
13245589 Group A 2mL
11414079 Group D 2mL
13215599 Group B 2mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 13295589 Supplier Thermo Scientific™ Supplier No. R30167401
  • ONLINE EXCLUSIVE PRICE
    £305.00
    £298.20 / 2mL
    Save £6.80
    2% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Used for serological identification of N. meningitidis by rapid slide agglutination and CIE procedures.

Remel Agglutinating Sera are used for serological identification of N. meningitidis by rapid slide agglutination and CIE procedures.

TRUSTED_SUSTAINABILITY

Specifications

Description Group W135
CE Marker Yes
Detectable Analytes N. meningitidis
Type Neisseria Meningitidis Test Kit
Organism Group Group W135
Target Organism Class Neisseria Meningitidis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.