Learn More
Thermo Scientific™ Remel™ Agglutinating Sera, Neisseria meningitidis
Used for serological identification of N. meningitidis by rapid slide agglutination and CIE procedures.
Brand:  Thermo Scientific™ R30166801
Code : 50
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 13245589
  • £347.00 / 2mL
In Stock
to see stock.
Description
Description
Remel Agglutinating Sera are used for serological identification of N. meningitidis by rapid slide agglutination and CIE procedures.
Specifications
Specifications
Neisseria meningitidis Group A
N. meningitidis
Group A
Yes
Neisseria Meningitidis Test Kit
Neisseria Meningitidis