Learn More
Thermo Scientific™ Remel™ Agglutinating Sera, E. coli Polyvalent
Product Code. 12627539
Change view
Click to view available options
Description:
E. coli Polyvalent 2
E. coli Polyvalent 3
E. coli Polyvalent 4
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Description Unit Size
12627539 E. coli Polyvalent 2 Each
12647539 E. coli Polyvalent 4 Each
12637539 E. coli Polyvalent 3 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 12627539 Supplier Thermo Scientific™ Supplier No. R30954901
  • ONLINE EXCLUSIVE PRICE
    £145.21
    £129.80 / Pack of 1
    Save £15.41
    10% Off
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
Used for serological identification of E. coli by slide and/or tube agglutination tests.
Remel Agglutinating Sera, E. coli Polyvalent is used for serological identification of E. coli by slide and/or tube agglutination tests.
TRUSTED_SUSTAINABILITY
Specifications
Description E. coli Polyvalent 2
Detectable Analytes E. coli
Type Agglutinating Sera
Packaging Type 2 mL/Vial
Quantity 2 mL
Specificity types O26, O55, O111, O119, O126