Learn More
Thermo Scientific™ Remel™ ADC Enrichment
Product Code. 11933262
Change view
Click to view available options
Quantity:
20 mL
Unit Size:
Pack of 10
Product Code. Quantity unitSize
11933262 20 mL Pack of 10
1 options
Product Code. 11933262 Supplier Thermo Scientific™ Supplier No. R450592
Remel ADC Enrichment is used in Middlebrook media.
Remel ADC Enrichment is used in Middlebrook media.
TRUSTED_SUSTAINABILITY
Specifications
Description Acetate Differential Agar (Sodium Acetate Agar)
Format D - slant
Product Type Enrichment
Quantity 20 mL