missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Remdesivir

Product Code. 16406677
Encompass_Preferred
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
This item is not returnable. View return policy

Product Code. 16406677

Brand: MedChemExpress HY104077

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Remdesivir (GS-5734), a nucleoside analogue with effective antiviral activity, has EC50s of 74 nM for SARS-CoV and MERS-CoV in HAE cells, and 30 nM for murine hepatitis virus in delayed brain tumor cells. Remdesivir is highly effective in the control of SARS-CoV-2 (COVID-19) infection in vitro.

Specifications

Molecular Weight (g/mol) 602.58 g/mol
Product Type Reference compound
Content And Storage Powder -20°C 3 years∣In solvent -80°C 6 months, -20°C 1 month
Shipping Condition Room temperature
Purity 98.3%
Quantity 5 mg
Solubility Information DMSO : 100 mg/mL (165.95 mM∣Need ultrasonic)
For Use With (Application) COVID-19-anti-virus
CAS 1809249-37-3
Synonym GS-5734
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.