missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Remdesivir
GREENER_CHOICE

Product Code. 18717556
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18717556

Brand: Tocris Bioscience™ 7226/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Viral RNA-dependent RNA polymerase (RdRP) inhibitor; broad spectrum antiviral nucelotide prodrug

Remdesivir is a viral RNA-dependent RNA polymerase inhibitor (RdRp) and broad spectrum antiviral nucleotide prodrug. Metabolized to active form, GS 441524 (Cat. No. 7227). Exhibits activity against a range of RNA viruses in vitro (EC50 values are 0.019, 0.07, 0.07 - 0.34, 0.086 - 0.14, and 23.15 μM against RSV, SARS-CoV, MERS-CoV, EBOV and SARS-CoV-2, respectively). Exhibits antiviral efficacy in animal models of MERS-CoV and EBOV infection.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Ethylbutyl (2 S)-2-[[-[[(2 R,3 S,4 R,5 R)-5-(4-aminopyrrolo(2,1-f)(1,2,4)triazin-7-yl)-5-cyano-3,4-dihydroxytetrahydrofuran-2-yl]methoxy]phenoxyphosphoryl]amino]propanoate
CAS 1809249-37-3
Quantity 10 mg
Target RNA/DNA Polymerase Inhibitors
Molecular Formula C27H35N6O8P
Purity 0.98
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.