missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ REM2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5103871
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total REM2.
Binds GTP saturably and exhibits a low intrinsic rate of GTP hydrolysis. Target protein is a Ras-related small GTP-binding protein that is found in high levels in neuronal tissues.
Specifications
| REM2 | |
| Polyclonal | |
| Unconjugated | |
| REM2 | |
| AW411893; GEM, REM, Kir family small G-protein; GTP-binding protein REM 2; GTP-binding protein REM2; rad and gem related GTP binding protein 2; rad and Gem-like GTP-binding protein 2; Rad and Gem-related 2 (rat homolog); RAS (RAD and GEM) like GTP binding 2; RAS (RAD and GEM)-like GTP binding 2; RAS-like GTP binding 2; Ras-related GTP-binding protein of the Rad/Gem/Kir family member 2; Ras-related GTP-binding protein of the Rad/Gem/Kir family, member 2; Rem2; RRAD and GEM like GTPase 2 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 140743, 161253, 64626 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q8IYK8, Q8VEL9, Q9WTY2 | |
| REM2 | |
| A synthesized peptide derived from human REM2(Accession Q8IYK8), corresponding to amino acid residues L48-S98. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction