missing translation for 'onlineSavingsMsg'
Learn More

Relugolix, MedChemExpress

Product Code. 16413107
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16413107

Brand: Medchem Express HY16474

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Relugolix (TAK-385) is a potent, orally active, nonpeptidic gonadotropin-releasing hormone (GnRH) antagonist. Relugolix possesses high affinity and potent antagonistic activity for human receptor (binding IC50=0.33 nM) and monkey receptor (IC50=0.32 nM) compared with TAK-013 (HY-100209). Relugolix is used for the study of sex-hormone-dependent diseases, such as including endometriosis, uterine fibroids and prostate cancer et al.

Relugolix (TAK-385) is a potent, orally active, nonpeptidic gonadotropin-releasing hormone (GnRH) antagonist. Relugolix possesses high affinity and potent antagonistic activity for human receptor (binding IC50=0.33 nM) and monkey receptor (IC50=0.32 nM) compared with TAK-013 (HY-100209). Relugolix is used for the study of sex-hormone-dependent diseases, such as including endometriosis, uterine fibroids and prostate cancer et al.

Specifications

Chemical Name or Material Relugolix
CAS 737789-87-6
Color Off-White
Purity Grade Notes Research
Molecular Formula C29H27F2N7O5S
Quantity 50 mg
Synonym TAK-385
Solubility Information DMSO : 100 mg/mL (160.35 mM; Need ultrasonic)
SMILES O=C(NOC)NC1=CC=C(C(S2)=C(CN(C)C)C(C(N3C4=NN=C(OC)C=C4)=O)=C2N(CC5=C(F)C=CC=C5F)C3=O)C=C1
Molecular Weight (g/mol) 623.63
Formula Weight 623.63
Percent Purity 99.51%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.